accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12024-24-7

GaGaSeSeSe

CAS RN 12024-24-7

IUPAC Name: digallium(3+) triselandiide
Synonyms: gallium(III) selenide
Molecular Weight (g/mol): 376.36
Molecular Formula: Ga2Se3
InChi Key: ZZEMEJKDTZOXOI-UHFFFAOYSA-N
SMILES: [Ga+3].[Ga+3].[Se--].[Se--].[Se--]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results