accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12022-95-6

SiFe

CAS RN 12022-95-6

IUPAC Name: silanidylidyneironylium
Synonyms: silanidylidyneironylium
Molecular Weight (g/mol): 83.93
Molecular Formula: FeSi
InChi Key: YSWDZWXHBYTMBR-UHFFFAOYSA-N
SMILES: [Si-]#[Fe+]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results