accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12009-21-1

OOOZrBa

CAS RN 12009-21-1

IUPAC Name: zirconium(4+) barium(2+) trioxidandiide
Synonyms: zirconium(4+) barium(2+) trioxidandiide
Molecular Weight (g/mol): 276.55
Molecular Formula: BaO3Zr
InChi Key: QLFKDQYBEKOEOT-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Zr+4].[Ba++]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results