accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12009-14-2

ONbOOONbOOBa

CAS RN 12009-14-2

IUPAC Name: diniobium(5+) dibarium(2+) heptaoxidandiide
Synonyms: diniobium(5+) dibarium(2+) heptaoxidandiide
Molecular Weight (g/mol): 572.46
Molecular Formula: Ba2Nb2O7
InChi Key: XEAHJNFQTFUOMO-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[Nb+5].[Nb+5].[Ba++].[Ba++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results