accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12007-25-9

BBMg

CAS RN 12007-25-9

IUPAC Name: diboron magnesium
Synonyms: magnesium diboride
Molecular Weight (g/mol): 45.93
Molecular Formula: B2Mg
InChi Key: PZKRHHZKOQZHIO-UHFFFAOYSA-N
SMILES: [B].[B].[Mg]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results