accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12004-78-3

AlAlAlTi

CAS RN 12004-78-3

IUPAC Name: trialuminium titanium
Synonyms: trialuminium titanium
Molecular Weight (g/mol): 128.81
Molecular Formula: Al3Ti
InChi Key: OQPDWFJSZHWILH-UHFFFAOYSA-N
SMILES: [Al].[Al].[Al].[Ti]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results