accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12004-04-5

OOOOOAlAlBaBa

CAS RN 12004-04-5

IUPAC Name: dialuminium(3+) dibarium(2+) pentaoxidandiide
Synonyms: dialuminium(3+) dibarium(2+) pentaoxidandiide
Molecular Weight (g/mol): 408.61
Molecular Formula: Al2Ba2O5
InChi Key: WIHSDLARTQXPRC-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[Al+3].[Al+3].[Ba++].[Ba++]
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)