accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1197193-30-8

H3COONNBr

CAS RN 1197193-30-8

IUPAC Name: ethyl 5-bromopyrimidine-2-carboxylate
Synonyms: ethyl 5-bromopyrimidine-2-carboxylate
Molecular Weight (g/mol): 231.05
Molecular Formula: C7H7BrN2O2
InChi Key: CZQFHUXIWZYQGP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NC=C(Br)C=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results