accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClBrCl

CAS RN 1193-72-2

IUPAC Name: 1-bromo-2,4-dichlorobenzene
Synonyms: 1-bromo-2,4-dichlorobenzene
Molecular Weight (g/mol): 225.89
Molecular Formula: C6H3BrCl2
InChi Key: ISHYFWKKWKXXPL-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Br)C(Cl)=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)