accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1193-12-0

CH3H3CHN

CAS RN 1193-12-0

IUPAC Name: 3,3-dimethylpiperidine
Synonyms: 3,3-dimethylpiperidine
Molecular Weight (g/mol): 113.2
Molecular Formula: C7H15N
InChi Key: CDODDZJCEADUQQ-UHFFFAOYSA-N
SMILES: CC1(C)CCCNC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results