accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNONH2OOOHHO

CAS RN 1187931-57-2

IUPAC Name: 1-(5-methyl-1,3,4-oxadiazol-2-yl)methanamine; oxalic acid
Synonyms: 1-(5-methyl-1,3,4-oxadiazol-2-yl)methanamine; oxalic acid
Molecular Weight (g/mol): 203.15
Molecular Formula: C6H9N3O5
InChi Key: BQNHEXDOLOGXGU-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.CC1=NN=C(CN)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results