CAS RN 1187188-59-5
CAS RN 1187188-59-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-[(1,3-Dihydro-1-hydroxy-2,1-benzoxaborol-5-yl)oxy]-benzamide, TRC
CAS: 1187188-59-5 Molecular Formula: C14H12BNO4 Molecular Weight (g/mol): 269.06 Synonym: Crisaborole Impurity IUPAC Name: 4-[(1-hydroxy-3H-2,1-benzoxaborol-5-yl)oxy]benzamide SMILES: NC(=O)c1ccc(Oc2ccc3B(O)OCc3c2)cc1