accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1186-31-8

CH3(S)CH3OHH2C

CAS RN 1186-31-8

IUPAC Name: (4S)-4-methylhept-1-en-4-ol
Synonyms: (4S)-4-methylhept-1-en-4-ol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: PYKQMQBNPLNLPS-MRVPVSSYSA-N
SMILES: CCC[C@](C)(O)CC=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results