accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1184-43-6

NNNNNNNNNNNNOOOOOOOOOOCH3H3CClCl

CAS RN 1184-43-6

IUPAC Name: 2-{4'-[3,5-bis(4-nitrophenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium-2-yl]-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl}-3,5-bis(4-nitrophenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium dichloride
Synonyms: 2-{4'-[3,5-bis(4-nitrophenyl)-1,2λ⁵,3,4-tetrazol-2-ylium-2-yl]-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl}-3,5-bis(4-nitrophenyl)-1,2λ⁵,3,4-tetrazol-2-ylium dichloride
Molecular Weight (g/mol): 907.64
Molecular Formula: C40H28Cl2N12O10
InChi Key: VCESGVLABVSDRO-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].COC1=CC(=CC=C1[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C(OC)=C1)[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O
Color 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results