accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 118289-07-9

HOFFF

CAS RN 118289-07-9

IUPAC Name: (2,4,6-trifluorophenyl)methanol
Synonyms: (2,4,6-trifluorophenyl)methanol
Molecular Weight (g/mol): 162.11
Molecular Formula: C7H5F3O
InChi Key: YJQDVBPZXNALEE-UHFFFAOYSA-N
SMILES: OCC1=C(F)C=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results