accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 118131-57-0

OOOOBaCF3CF3CF3CF3

CAS RN 118131-57-0

IUPAC Name: barium(2+) 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate
Synonyms: barium(2+) 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate
Molecular Weight (g/mol): 344.38
Molecular Formula: C5HBaF6O2
InChi Key: KVBKEWTWOCKTBS-UHFFFAOYSA-M
SMILES: [Ba++].[O-]C(=CC(=O)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results