accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 118101-09-0

H3CNONHONHCH3N

CAS RN 118101-09-0

IUPAC Name: 1-(1-methyl-2-oxo-5-phenyl-2,3-dihydro-1H-1,4-benzodiazepin-3-yl)-3-(3-methylphenyl)urea
Synonyms: 1-(1-methyl-2-oxo-5-phenyl-3H-1,4-benzodiazepin-3-yl)-3-(3-methylphenyl)urea
Molecular Weight (g/mol): 398.47
Molecular Formula: C24H22N4O2
InChi Key: KDFQABSFVYLGPM-UHFFFAOYNA-N
SMILES: CN1C2=CC=CC=C2C(=NC(NC(=O)NC2=CC=CC(C)=C2)C1=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results