accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SFFFFFBrOOH

CAS RN 1180675-98-2

IUPAC Name: 3-bromo-5-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Synonyms: 3-bromo-5-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Molecular Weight (g/mol): 327.06
Molecular Formula: C7H4BrF5O2S
InChi Key: GGDWTFSLWQRHPR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC(Br)=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results