accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNIBr

CAS RN 118000-66-1

IUPAC Name: 2-(4-bromophenyl)-6-iodoimidazo[1,2-a]pyridine
Synonyms: 2-(4-bromophenyl)-6-iodoimidazo[1,2-a]pyridine
Molecular Weight (g/mol): 399.03
Molecular Formula: C13H8BrIN2
InChi Key: DOUYIPIXURZQRX-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CN2C=C(I)C=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results