accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1180-71-8

CH3CH3O(S)OO(R)(R)(S)H3C(S)OO(S)O(R)(R)CH3O(R)O

CAS RN 1180-71-8

IUPAC Name: (1R,2R,7S,10R,13R,14R,16S,19S,20S)-19-(furan-3-yl)-9,9,13,20-tetramethyl-4,8,15,18-tetraoxahexacyclo[11.9.0.0²,⁷.0²,¹⁰.0¹⁴,¹⁶.0¹⁴,²⁰]docosane-5,12,17-trione
Synonyms: limonin obaculactone evodin limonine evodia fruit citrolimonin di-delta-lactone limonoic acid dictamnolactone
Molecular Weight (g/mol): 470.52
Molecular Formula: C26H30O8
InChi Key: KBDSLGBFQAGHBE-MSGMIQHVSA-N
SMILES: CC1(C)O[C@H]2CC(=O)OC[C@@]22[C@H]1CC(=O)[C@]1(C)[C@@H]2CC[C@@]2(C)[C@@H](OC(=O)[C@H]3O[C@@]123)C1=COC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (2)