CAS RN 1174161-86-4
CAS RN 1174161-86-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
SCR-1481B1, MedChemExpress
MedChemExpress SCR-1481B1 (c-Met inhibitor 2) is a potent compound that has activity against cancers dependent upon Met activation and also has activity against cancers as a VEGFR inhibitor.