accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 116405-43-7

NPPOOFFFPhPhPhPhPhPh

CAS RN 116405-43-7

IUPAC Name: triphenyl[(triphenyl-λ⁵-phosphanylidene)amino]phosphanium trifluoroacetate
Synonyms: triphenyl[(triphenyl-λ⁵-phosphanylidene)amino]phosphanium trifluoroacetate
Molecular Weight (g/mol): 651.61
Molecular Formula: C38H30F3NO2P2
InChi Key: NDLHFNFVQBXEPE-UHFFFAOYSA-M
SMILES: [O-]C(=O)C(F)(F)F.C1=CC=C(C=C1)[P+](N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results