CAS RN 116173-67-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
beta-Alanine-2,2,3,3-d4, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
Beta-Alanine-2,2,3,3-D4, TRC
CAS: 116173-67-2 Molecular Formula: C3 D4 H3 N O2 Molecular Weight (g/mol): 93.118 Synonym: β-Alanine-2,2,3,3-d4 (9CI, ACI),β-Alanine-d4,β-Alanine-2,2,3,3-D4,3-Aminopropanoic-2,2,3,3-d4 acid,3-Aminopropionic-d4 acid IUPAC Name: 3-amino-2,2,3,3-tetradeuteriopropanoic acid SMILES: [2H]C([2H])(N)C([2H])([2H])C(=O)O