accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 116070-39-4

CH3OFFF

CAS RN 116070-39-4

IUPAC Name: 3-methyl-5-(trifluoromethyl)benzaldehyde
Synonyms: 3-methyl-5-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 188.15
Molecular Formula: C9H7F3O
InChi Key: KFRRYUVXFZQWGS-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C=O)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results