CAS RN 115730-24-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
ZT 52656A hydrochloride, MedChemExpress
MedChemExpress ZT 52656A is a selective kappa opioid agonist, used for the prevention or alleviation of pain in the eye.