accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 115199-84-3

ClCH3N(S)NOClClH

CAS RN 115199-84-3

IUPAC Name: hydrogen 2-(3,4-dichlorophenyl)-N-methyl-N-[(1S)-1-phenyl-2-(pyrrolidin-1-yl)ethyl]acetamide chloride
Synonyms: hydrogen 2-(3,4-dichlorophenyl)-N-methyl-N-[(1S)-1-phenyl-2-(pyrrolidin-1-yl)ethyl]acetamide chloride
Molecular Weight (g/mol): 427.79
Molecular Formula: C21H25Cl3N2O
InChi Key: VFLWVWZSDBTGQJ-VEIFNGETSA-N
SMILES: [H+].[Cl-].CN([C@H](CN1CCCC1)C1=CC=CC=C1)C(=O)CC1=CC=C(Cl)C(Cl)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results