accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 114873-06-2

CH3(S)HNOOCH3H3CCH3OHO

CAS RN 114873-06-2

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(3-methylphenyl)propanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(3-methylphenyl)propanoic acid
Molecular Weight (g/mol): 279.34
Molecular Formula: C15H21NO4
InChi Key: HBBXWMALJNZDOM-LBPRGKRZSA-N
SMILES: CC1=CC=CC(C[C@H](NC(=O)OC(C)(C)C)C(O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)