accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 114476-84-5

ClOOCl

CAS RN 114476-84-5

IUPAC Name: 2-(3-chlorophenoxy)acetyl chloride
Synonyms: 2-(3-chlorophenoxy)acetyl chloride
Molecular Weight (g/mol): 205.03
Molecular Formula: C8H6Cl2O2
InChi Key: KPINZNVDHXIYAR-UHFFFAOYSA-N
SMILES: ClC(=O)COC1=CC(Cl)=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)