CAS RN 1141934-97-5
CAS RN 1141934-97-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Emixustat hydrochloride, MedChemExpress
MedChemExpress Emixustat hydrochloride strongly inhibits 11-cis-retinol production with IC50 values of 232 (plusmn) 3 nM.