accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 113760-29-5

CH3OSNOCH3(E/Z)

CAS RN 113760-29-5

IUPAC Name: 6,6-dimethyl-2-(phenylimino)-4,5,6,7-tetrahydro-2H-1,3-benzoxathiol-4-one
Synonyms: 6,6-dimethyl-2-(phenylimino)-5,7-dihydro-1,3-benzoxathiol-4-one
Molecular Weight (g/mol): 273.35
Molecular Formula: C15H15NO2S
InChi Key: XFSSJIQCIPSBHJ-UHFFFAOYSA-N
SMILES: CC1(C)CC2=C(SC(O2)=NC2=CC=CC=C2)C(=O)C1
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results