accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1135242-13-5

CH3OONHNNHOH3CHCl

CAS RN 1135242-13-5

IUPAC Name: ethyl 4-({2-[(2-methylphenyl)formamido]ethyl}amino)piperidine-1-carboxylate hydrochloride
Synonyms: ethyl 4-({2-[(2-methylphenyl)formamido]ethyl}amino)piperidine-1-carboxylate hydrochloride
Molecular Weight (g/mol): 369.89
Molecular Formula: C18H28ClN3O3
InChi Key: XKJQVUIXSBOCPP-UHFFFAOYSA-N
SMILES: Cl.CCOC(=O)N1CCC(CC1)NCCNC(=O)C1=CC=CC=C1C
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results