accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 113168-57-3

NONNHClHCl

CAS RN 113168-57-3

IUPAC Name: 1-phenyl-3,3-bis[(pyridin-4-yl)methyl]-2,3-dihydro-1H-indol-2-one dihydrochloride
Synonyms: linopirdine dihydrochloride
Molecular Weight (g/mol): 464.39
Molecular Formula: C26H23Cl2N3O
InChi Key: ZEVVHCGTTNRYOY-UHFFFAOYSA-N
SMILES: Cl.Cl.O=C1N(C2=CC=CC=C2C1(CC1=CC=NC=C1)CC1=CC=NC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results