CAS RN 113137-29-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(4-Chloro-6-methoxyphenyl)-2,2-dimethylpropanamide, TRC
CAS: 113137-29-4 Molecular Formula: C12 H16 Cl N O2 Molecular Weight (g/mol): 241.71 IUPAC Name: N-(4-chloro-2-methoxyphenyl)-2,2-dimethylpropanamide SMILES: COc1cc(Cl)ccc1NC(=O)C(C)(C)C