CAS RN 113-53-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dothiepin, TRC
CAS: 113-53-1 Molecular Formula: C19H21NS Molecular Weight (g/mol): 295.44 Synonym: 1-Propanamine, 3-dibenzo[b,e]thiepin-11(6H)-ylidene-N,N-dimethyl-,Dibenzo[b,e]thiepin-δ11(6H),γ-propylamine, N,N-dimethyl- (7CI,8CI),3-Dibenzo[b,e]thiepin-11(6H)-ylidene-N,N-dimethyl-1-propanamine,Dibenzo[b,e]thiepin, 1-propanamine deriv.,Dosulepin,Dosulepine,Dothep,Dothiepin,IZ 914,Prothiaden,Prothiadene SMILES: CN(C)CC\C=C\1/c2ccccc2CSc3ccccc13