accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1129-37-9

OHNNOO(E)

CAS RN 1129-37-9

IUPAC Name: (E)-N-[(4-nitrophenyl)methylidene]hydroxylamine
Synonyms: P-nitrobenzaldoxime
Molecular Weight (g/mol): 166.14
Molecular Formula: C7H6N2O3
InChi Key: WTLPAVBACRIHHC-VMPITWQZSA-N
SMILES: O\N=C\C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results