accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 112741-49-8

IUPAC Name: tert-butyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate
Synonyms: tert-butyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate
Molecular Weight (g/mol): 353.42
Molecular Formula: C21H23NO4
InChi Key: MRUKRSQUUNYOFK-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC(=O)OC(C1C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results