accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1125-88-8

CH3OOCH3

CAS RN 1125-88-8

IUPAC Name: (dimethoxymethyl)benzene
Synonyms: α,α-dimethoxytoluene benzaldehyde dimethylacetal
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: HEVMDQBCAHEHDY-UHFFFAOYSA-N
SMILES: COC(OC)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (7)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (4)
  • (3)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (7)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (4)
  • (5)