accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1122-54-9

H3CON

CAS RN 1122-54-9

IUPAC Name: 1-(pyridin-4-yl)ethan-1-one
Synonyms: 4-acetylpyridine
Molecular Weight (g/mol): 121.14
Molecular Formula: C7H7NO
InChi Key: WMQUKDQWMMOHSA-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)