accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1122-43-6

CH3OHH3CN

CAS RN 1122-43-6

IUPAC Name: 2,6-dimethylpyridin-3-ol
Synonyms: 2,6-dimethyl-3-oxypyridine
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: AMQJAKKATFMFTR-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C(C)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results