CAS RN 1120-57-6
CAS RN 1120-57-6
IUPAC Name:
chlorocyclobutane
Synonyms:
chlorocyclobutane
Molecular Weight (g/mol):
90.55
Molecular Formula:
C4H7Cl
InChi Key:
STJYMUBZVMSMBP-UHFFFAOYSA-N
SMILES:
ClC1CCC1
1
–
1
of
1
results
1
Chlorocyclobutane, 97%
CAS: 1120-57-6 Molecular Formula: C4H7Cl Molecular Weight (g/mol): 90.55 MDL Number: MFCD00013736 InChI Key: STJYMUBZVMSMBP-UHFFFAOYSA-N Synonym: cyclobutyl chloride,cyclobutane, chloro,chloro-cyclobutane,cyclobutylchloride,chloranylcyclobutane,1-chlorocyclobutane,acmc-1butg,chlorocyclobutane, 97+% PubChem CID: 70712 IUPAC Name: chlorocyclobutane SMILES: C1CC(C1)Cl