accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NH2CH3O(Z)

CAS RN 1118-66-7

IUPAC Name: (3Z)-4-aminopent-3-en-2-one
Synonyms: (Z)-4-aminopent-3-en-2-one
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: OSLAYKKXCYSJSF-ARJAWSKDSA-N
SMILES: C\C(N)=C\C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results