accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 111781-49-8

CH3NNFF

CAS RN 111781-49-8

IUPAC Name: 2-(1,1-difluoroethyl)pyrazine
Synonyms: 2-(1,1-difluoroethyl)pyrazine
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H6F2N2
InChi Key: OXKFWGSACYGCSY-UHFFFAOYSA-N
SMILES: CC(F)(F)C1=NC=CN=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)