accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 111735-27-4

H3COON

CAS RN 111735-27-4

IUPAC Name: N-benzyl-4-methoxy-N-phenylbenzamide
Synonyms: N-benzyl-4-methoxy-N-phenylbenzamide
Molecular Weight (g/mol): 317.39
Molecular Formula: C21H19NO2
InChi Key: BLUMCNOTEFEZGV-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)N(CC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results