CAS RN 111149-90-7
CAS RN 111149-90-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lodelaben, MedChemExpress
MedChemExpress Lodelaben is a human neutrophil elastase inhibitor with an IC50 and Ki of 0.5 and 1.5 μM, respectively.