CAS RN 1109276-89-2
CAS RN 1109276-89-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ PF 04620110
Potent and selective DGAT-1 inhibitor; orally bioavailable
2
PF-04620110, MedChemExpress
MedChemExpress PF-04620110 is a potent, selective and orally bioavailable diglyceride acyltransferase-1 (DGAT-1) inhibitor with an IC50 of 19 nM.