accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 11092-32-3

OAlOAlO

CAS RN 11092-32-3

IUPAC Name: oxo[(oxoalumanyl)oxy]alumane
Synonyms: sapphire powder
Molecular Weight (g/mol): 101.96
Molecular Formula: Al2O3
InChi Key: TWNQGVIAIRXVLR-UHFFFAOYSA-N
SMILES: O=[Al]O[Al]=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results