accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 110-67-8

H3CON

CAS RN 110-67-8

IUPAC Name: 3-methoxypropanenitrile
Synonyms: propanenitrile, 3-methoxy-
Molecular Weight (g/mol): 85.11
Molecular Formula: C4H7NO
InChi Key: OOWFYDWAMOKVSF-UHFFFAOYSA-N
SMILES: COCCC#N
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)