CAS RN 1093679-73-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Caffeic Acid 3-Beta-D-Glucuronide, TRC
CAS: 1093679-73-2 Molecular Formula: C15H16O10 Molecular Weight (g/mol): 356.28 Synonym: 5-[(1E)-2-Carboxyethenyl]-2-hydroxyphenyl β-D-Glucopyranosiduronic Acid; IUPAC Name: (2S,3S,4S,5R,6S)-6-[5-[(E)-2-carboxyethenyl]-2-hydroxyphenoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid SMILES: C1=CC(=C(C=C1C=CC(=O)O)OC2C(C(C(C(O2)C(=O)O)O)O)O)O