CAS RN 1092461-37-4
CAS RN 1092461-37-4
IUPAC Name:
1-[3-bromo-5-(trifluoromethoxy)phenyl]methanamine
Synonyms:
1-[3-bromo-5-(trifluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol):
270.05
Molecular Formula:
C8H7BrF3NO
InChi Key:
LYWSEMAXLWKKLQ-UHFFFAOYSA-N
SMILES:
NCC1=CC(OC(F)(F)F)=CC(Br)=C1
1
–
1
of
1
results
1
3-Bromo-5-(trifluoromethoxy)benzylamine, 97%, Thermo Scientific™
CAS: 1092461-37-4 Molecular Formula: C8H7BrF3NO Molecular Weight (g/mol): 270.049 MDL Number: MFCD11519380 InChI Key: LYWSEMAXLWKKLQ-UHFFFAOYSA-N Synonym: 3-bromo-5-trifluoromethoxy benzylamine,1-3-bromo-5-trifluoromethoxy phenyl methanamine,3-bromo-5-trifluoromethoxy benzylamine, jrd PubChem CID: 53414437 IUPAC Name: [3-bromo-5-(trifluoromethoxy)phenyl]methanamine SMILES: C1=C(C=C(C=C1OC(F)(F)F)Br)CN