CAS RN 1092461-34-1
CAS RN 1092461-34-1
IUPAC Name:
2-[3-bromo-5-(trifluoromethoxy)phenyl]acetonitrile
Synonyms:
2-[3-bromo-5-(trifluoromethoxy)phenyl]acetonitrile
Molecular Weight (g/mol):
280.04
Molecular Formula:
C9H5BrF3NO
InChi Key:
WWIWPXWPBQBQIO-UHFFFAOYSA-N
SMILES:
FC(F)(F)OC1=CC(Br)=CC(CC#N)=C1
1
–
1
of
1
results
1
3-Bromo-5-(trifluoromethoxy)phenylacetonitrile, JRD, 97%, Thermo Scientific™
CAS: 1092461-34-1 Molecular Formula: C9H5BrF3NO Molecular Weight (g/mol): 280.04 MDL Number: MFCD11519377 InChI Key: WWIWPXWPBQBQIO-UHFFFAOYSA-N Synonym: 3-bromo-5-trifluoromethoxy phenylacetonitrile,2-3-bromo-5-trifluoromethoxy phenyl acetonitrile,3-bromo-5-trifluoromethoxy phenylacetonitrile, jrd PubChem CID: 54772377 IUPAC Name: 2-[3-bromo-5-(trifluoromethoxy)phenyl]acetonitrile SMILES: FC(F)(F)OC1=CC(Br)=CC(CC#N)=C1